vibrant-m

R2R Monolayer large grain CVD graphene on silicon wafer

vibrant-m
Empirical Formula (Hill Notation):
C
CAS号:
分子量:
12.01

产品对比指南

请使用如下性质配置您的对比表。(最多可选择3。)

Select Attribute
Select Attribute
Select Attribute
Sort By: Default
货号
产品描述
价格
R2R Monolayer large grain CVD graphene on silicon wafer, 4 in diameter
R2R Monolayer large grain CVD graphene on silicon wafer, 6 in diameter