Merck
山梨醇- CAS 50-70-4 - Calbiochem A sugar alcohol produced by reduction of glucose.

山梨醇- CAS 50-70-4 - Calbiochem

山梨醇- CAS 50-70-4 - Calbiochem A sugar alcohol produced by reduction of glucose.
Synonyms:
D -山梨醇, D-山梨糖醇
Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
山梨醇- CAS 50-70-4 - Calbiochem, A sugar alcohol produced by reduction of glucose.