Merck
四氢呋喃EMPROVE ® ESSENTIAL tetrahydrofuran processed under GMP conditions for use in pharmaceutical production, EMPROVE® ESSENTIAL

四氢呋喃EMPROVE ® ESSENTIAL

四氢呋喃EMPROVE ® ESSENTIAL tetrahydrofuran processed under GMP conditions for use in pharmaceutical production, EMPROVE® ESSENTIAL
Synonyms:
THF, 环氧丁烷, 环氧丙烷, 四氢呋喃
Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
四氢呋喃EMPROVE ® ESSENTIAL, tetrahydrofuran processed under GMP conditions for use in pharmaceutical production, EMPROVE® ESSENTIAL