• 主页
  • 查找结果
  • Use of a copper-chelated lipid speeds up NMR measurements from membrane proteins.
Use of a copper-chelated lipid speeds up NMR measurements from membrane proteins.
Journal of the American Chemical Society (2010-05-04)
Kazutoshi Yamamoto, Jiadi Xu, Karen E Kawulka, John C Vederas, Ayyalusamy Ramamoorthy
摘要
Recent studies have demonstrated the abilities of solid-state NMR techniques to solve atomic-level-resolution structures and dynamics of membrane-associated proteins and peptides. However, high-throughput applications of solid-state NMR spectroscopy are hampered by long acquisition times due to the low sensitivity of the technique. In this study, we demonstrate the use of a paramagnetic copper-chelated lipid to enhance the spin-lattice relaxation and thereby speed up solid-state NMR measurements. Fluid lamellar-phase bicelles composed of a lipid, detergent, and the copper-chelated lipid and containing a uniformly (15)N-labeled antimicrobial peptide, subtilosin A, were used at room temperature. The use of a chelating lipid reduces the concentration of free copper and limits RF-induced heating, a major problem for fluid samples. Our results demonstrate a 6.2-fold speed increase and a 2.7-fold improvement in signal-to-noise ratio for solid-state NMR experiments under magic-angle spinning and static conditions, respectively. Furthermore, solid-state NMR measurements are shown to be feasible even for nanomole concentrations of a membrane-associated peptide.
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。