HK
ZH
应用
产品
服务
支持
Home
Search Results
Intracellular BH3 Profiling Reveals Shifts in Antiapoptotic Dependency in Human B Cell Maturation and Mitogen-Stimulated Proliferation
Intracellular BH3 Profiling Reveals Shifts in Antiapoptotic Dependency in Human B Cell Maturation and Mitogen-Stimulated Proliferation
Journal of Immunology (2018)
Dai J and Luftig MA