• 主页
  • 查找结果
  • Signal Integration of IFN-I and IFN-II With TLR4 Involves Sequential Recruitment of STAT1-Complexes and NFκB to Enhance Pro-inflammatory Transcription.
Signal Integration of IFN-I and IFN-II With TLR4 Involves Sequential Recruitment of STAT1-Complexes and NFκB to Enhance Pro-inflammatory Transcription.
Frontiers in immunology (2019-06-25)
Anna Piaszyk-Borychowska, Lajos Széles, Attila Csermely, Hsin-Chien Chiang, Joanna Wesoły, Chien-Kuo Lee, Laszlo Nagy, Hans A R Bluyssen
摘要
Atherosclerosis is a chronic inflammatory disease of the blood vessels, characterized by atherosclerotic lesion formation. Vascular Smooth Muscle Cells (VSMC), macrophages (MΦ), and dendritic cells (DC) play a crucial role in vascular inflammation and atherosclerosis. Interferon (IFN)α, IFNγ, and Toll-like receptor (TLR)4 activate pro-inflammatory gene expression and are pro-atherogenic. Gene expression regulation of many pro-inflammatory genes has shown to rely on Signal Integration (SI) between IFNs and TLR4 through combinatorial actions of the Signal Transducer and Activator of Transcription (STAT)1 complexes ISGF3 and γ-activated factor (GAF), and Nuclear Factor-κB (NFκB). Thus, IFN pre-treatment ("priming") followed by LPS stimulation leads to enhanced transcriptional responses as compared to the individual stimuli. To characterize the mechanism of priming-induced IFNα + LPS- and IFNγ + LPS-dependent SI in vascular cells as compared to immune cells, we performed a comprehensive genome-wide analysis of mouse VSMC, MΦ, and DC in response to IFNα, IFNγ, and/or LPS. Thus, we identified IFNα + LPS or IFNγ + LPS induced genes commonly expressed in these cell types that bound STAT1 and p65 at comparable γ-activated sequence (GAS), Interferon-stimulated response element (ISRE), or NFκB sites in promoter proximal and distal regions. Comparison of the relatively high number of overlapping ISRE sites in these genes unraveled a novel role of ISGF3 and possibly STAT1/IRF9 in IFNγ responses. In addition, similar STAT1-p65 co-binding modes were detected for IFNα + LPS and IFNγ + LPS up-regulated genes, which involved recruitment of STAT1 complexes preceding p65 to closely located GAS/NFκB or ISRE/NFκB composite sites already upon IFNα or IFNγ treatment. This STAT1-p65 co-binding significantly increased after subsequent LPS exposure and correlated with histone acetylation, PolII recruitment, and amplified target gene transcription in a STAT1-p65 co-bound dependent manner. Thus, co-binding of STAT1-containing transcription factor complexes and NFκB, activated by IFN-I or IFN-II together with LPS, provides a platform for robust transcriptional activation of pro-inflammatory genes. Moreover, our data offer an explanation for the comparable effects of IFNα or IFNγ priming on TLR4-induced activation in vascular and immune cells, with important implications in atherosclerosis.
材料
货号
品牌
产品描述
Sigma-Aldrich
氯化钠, ACS reagent, ≥99.0%
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
碳酸氢钠, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
抗生素抗霉菌溶液(100×),稳定, with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
甘氨酸, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
脂多糖 来源于大肠杆菌 0111:B4, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
SAFC
RPMI-1640 培养基, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
抗小鼠IgG(全分子)-过氧化物酶 兔抗, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
苯甲磺酰氟 溶液, ~0.1 M in ethanol (T)
Sigma-Aldrich
弹性蛋白酶 来源于猪胰腺, Type I, ≥4.0 units/mg protein
Sigma-Aldrich
抗-兔IgG(全分子)-过氧化物酶 山羊抗, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
S-亚硝基-N-乙酰基-DL-青霉胺, ≥97%, powder
Sigma-Aldrich
双琥珀酰亚胺戊二酸酯, ≥97.0% (CHN)
Sigma-Aldrich
抗RNA聚合酶II抗体,克隆CTD4H8, clone CTD4H8, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho-STAT2 (Tyr689) Antibody, Upstate®, from rabbit