• 主页
  • 查找结果
  • Environmental and human relevant PFOS and PFOA doses alter human mesenchymal stem cell self-renewal, adipogenesis and osteogenesis.
Environmental and human relevant PFOS and PFOA doses alter human mesenchymal stem cell self-renewal, adipogenesis and osteogenesis.
Ecotoxicology and environmental safety (2018-11-27)
Shuyu Liu, Renjun Yang, Nuoya Yin, Yuan-Liang Wang, Francesco Faiola
摘要
PFOS and PFOA are two of the most abundant perfluorinated compounds (PFCs) in the environment. Previous studies have reported they have a long half-life (up to five years) once they enter into the human body. Moreover, they can potentially promote the adipogenic process by activating PPARγ. However, little is known about PFOS and PFOA chronic health impacts on humans. In this study, we employed primary human mesenchymal stem cells (hMSCs) and demonstrated that PFOS and PFOA exerted acute cytotoxicity and affected adipogenesis and osteogenesis at environmental and human relevant doses. In fact, PFOS and PFOA impaired the proper expression of CD90 (a surface antigen highly enriched in undifferentiated hMSCs) and promoted adipogenesis, presumably via their interaction with PPARγ. Moreover, PFOA partly disturbed osteogenesis. Thus, our findings not only validated the health risks of PFOS and PFOA, but also revealed new potential long-term PFOS/PFOA impacts on humans.
材料
货号
品牌
产品描述
Sigma-Aldrich
2',7'-二氯荧光素二乙酸酯, ≥97%
Sigma-Aldrich
抗坏血酸磷酸酯镁 倍半镁盐 水合物, ≥95%
Sigma-Aldrich
β-甘油磷酸盐 二钠盐 水合物, BioUltra, suitable for cell culture, suitable for plant cell culture, ≥99% (titration)
Sigma-Aldrich
L -抗坏血酸, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
全氟辛酸, 95%
Sigma-Aldrich
亚油酸, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
尼罗红, Technical grade
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, low endotoxin, lyophilized powder, Cohn Analog, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
全氟辛基磺酸 钾盐, ≥98.0% (T)
Sigma-Aldrich
八乙二醇, ≥95% (oligomer purity)