Merck
  • 主页
  • 查找结果
  • Environmental Oxygen Exposure Allows for the Evolution of Interdigital Cell Death in Limb Patterning.
Environmental Oxygen Exposure Allows for the Evolution of Interdigital Cell Death in Limb Patterning.
Developmental cell (2019-06-18)
Ingrid Rosenburg Cordeiro, Kaori Kabashima, Haruki Ochi, Keijiro Munakata, Chika Nishimori, Mara Laslo, James Hanken, Mikiko Tanaka
摘要
Amphibians form fingers without webbing by differential growth between digital and interdigital regions. Amniotes, however, employ interdigital cell death (ICD), an additional mechanism that contributes to a greater variation of limb shapes. Here, we investigate the role of environmental oxygen in the evolution of ICD in tetrapods. While cell death is restricted to the limb margin in amphibians with aquatic tadpoles, Eleutherodactylus coqui, a frog with terrestrial-direct-developing eggs, has cell death in the interdigital region. Chicken requires sufficient oxygen and reactive oxygen species to induce cell death, with the oxygen tension profile itself being distinct between the limbs of chicken and Xenopus laevis frogs. Notably, increasing blood vessel density in X. laevis limbs, as well as incubating tadpoles under high oxygen levels, induces ICD. We propose that the oxygen available to terrestrial eggs was an ecological feature crucial for the evolution of ICD, made possible by conserved autopod-patterning mechanisms.