HK
ZH
应用
产品
服务
支持
Home
Search Results
Cysteine cathepsins: from structure, function and regulation to new frontiers
Cysteine cathepsins: from structure, function and regulation to new frontiers
Biochimica et Biophysica Acta (BBA)-Proteins and Proteomics (2012)
Turk V, et al