  • Search Results
  • Impairment of neocortical long-term potentiation in mice deficient of endothelial nitric oxide synthase.
Impairment of neocortical long-term potentiation in mice deficient of endothelial nitric oxide synthase.
Journal of neurophysiology (1999-02-26)
S Haul, A Gödecke, J Schrader, H L Haas, H J Luhmann
ABSTRACT
The role of the possible retrograde messenger nitric oxide (NO) in the induction of long-term potentiation (LTP) was studied in supragranular layers of somatosensory cortical slices obtained from adult mice. High-frequency stimulation produced a slowly rising, long-lasting (50 min) and significant (P < 0.001) increase in the extracellular synaptic response by 23%. The induction of LTP was independent from activation of N-methyl-D-aspartate (NMDA) receptors, but prevented by bath application of NG-nitro-L-arginine methyl ester (L-NAME), indicating that one or several of the different NO synthases (NOS) produced NO within the postsynaptic neuron. No LTP could be induced in knockout mice lacking the endothelial NOS (eNOS) isoform. These data suggest that eNOS is involved in an NMDA receptor-independent form of LTP in the rodent cerebral cortex.
Sigma-Aldrich
Nω-硝基-L-精氨酸甲酯 盐酸盐, ≥98% (TLC), powder