• 主页
  • 查找结果
  • Mechanistic investigations into the encapsulation and release of small molecules and proteins from a supramolecular nucleoside gel in vitro and in vivo.
Mechanistic investigations into the encapsulation and release of small molecules and proteins from a supramolecular nucleoside gel in vitro and in vivo.
Journal of controlled release : official journal of the Controlled Release Society (2019-11-05)
Maria Galini Faidra Angelerou, Robert Markus, Vasiliki Paraskevopoulou, Rugerro Foralosso, Philip Clarke, Clara V Alvarez, Miguel Chenlo, Litty Johnson, Catrin Rutland, Stephanie Allen, Christopher Brasnett, Annela Seddon, Mischa Zelzer, Maria Marlow
摘要
Supramolecular gels have recently emerged as promising biomaterials for the delivery of a wide range of bioactive molecules, from small hydrophobic drugs to large biomolecules such as proteins. Although it has been demonstrated that each encapsulated molecule has a different release profile from the hydrogel, so far diffusion and steric impediment have been identified as the only mechanisms for the release of molecules from supramolecular gels. Erosion of a supramolecular gel has not yet been reported to contribute to the release profiles of encapsulated molecules. Here, we use a novel nucleoside-based supramolecular gel as a drug delivery system for proteins with different properties and a hydrophobic dye and describe for the first time how these materials interact, encapsulate and eventually release bioactive molecules through an erosion-based process. Through fluorescence microscopy and spectroscopy as well as small angle X-ray scattering, we show that the encapsulated molecules directly interact with the hydrogel fibres - rather than being physically entrapped in the gel network. The ability of these materials to protect proteins against enzymatic degradation is also demonstrated here for the first time. In addition, the released proteins were proven to be functional in vitro. Real-time fluorescence microscopy together with macroscopic release studies confirm that erosion is the key release mechanism. In vivo, the gel completely degrades after two weeks and no signs of inflammation are detected, demonstrating its in vivo safety. By establishing the contribution of erosion as a key driving force behind the release of bioactive molecules from supramolecular gels, this work provides mechanistic insight into the way molecules with different properties are encapsulated and released from a nucleoside-based supramolecular gel and sets the basis for the design of more tailored supramolecular gels for drug delivery applications.
材料
货号
品牌
产品描述
Sigma-Aldrich
杜氏磷酸盐缓冲盐水, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
溶菌酶 来源于鸡蛋白, lyophilized powder, protein ≥90 %, ≥40,000 units/mg protein
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, lyophilized powder, ≥2,500 units/mg protein (E1%/280)
Sigma-Aldrich
α-糜蛋白酶 来源于牛胰腺, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
2'-脱氧胞苷, ≥99% (HPLC)
Sigma-Aldrich
β-乳球蛋白 B 来源于牛奶, ≥90% (PAGE)
Sigma-Aldrich
1,1′-二十八烷基-3,3,3′,3′-四甲基吲哚菁高氯酸盐, 97%