  • Search Results
  • DDAH-2 alleviates contrast medium iopromide-induced acute kidney injury through nitric oxide synthase.
DDAH-2 alleviates contrast medium iopromide-induced acute kidney injury through nitric oxide synthase.
Clinical science (London, England : 1979) (2019-11-26)
Heng-Huei Lin, Tzong-Shyuan Lee, Shing-Jong Lin, Yi-Chen Yeh, Tse-Min Lu, Chiao-Po Hsu
ABSTRACT
Contrast medium-induced acute kidney injury (CI-AKI) is one of the most common causes of hospital-acquired acute renal failure. However, the pathogenesis of CI-AKI remains unclear. Asymmetric dimethylarginine (ADMA) is an endogenous nitric oxide synthase (NOS) inhibitor that is largely metabolised by dimethylarginine dimethylaminohydroxylase (DDAH) in humans. Two isoforms of DDAH exist, namely, DDAH-1 and DDAH-2. In the present study, we examined whether the DDAH-2/ADMA/NOS pathway is involved in the pathogenesis of CI-AKI. Exposure to the contrast medium iopromide led to increase in creatinine and blood urea nitrogen (BUN) levels, accumulation of ADMA, increase in reactive oxygen species (ROS) generation, and an inflammatory response in mice kidney tissue. The injection of adenovirus-harbouring DDAH-2 lowered renal ADMA levels and had a reno-protective effect against contrast-medium injury by decreasing cell apoptosis, ROS, and fibrosis. By contrast, contrast medium-induced renal injury was exacerbated in heterozygous DDAH-2 knockout mice. In the in vitro study, overexpression of DDAH-2 increased the levels of nitrite and intracellular cGMP, while the DDAH-2 knockdown induced the opposite effect. These findings were also observed in the in vivo sample. Our findings provide the first evidence that the DDAH-2/ADMA/NOS pathway is involved in the pathogenesis of CI-AKI and that the protective effect of DDAH-2 probably arises from the modulation of NOS activity, oxidative stress, and the inflammatory process.
Sigma-Aldrich
N-乙酰基-L-半胱氨酸, BioReagent, suitable for cell culture
Sigma-Aldrich
单克隆抗 β-肌动蛋白抗体 小鼠抗, clone AC-15, ascites fluid
Sigma-Aldrich
Nω-硝基-L-精氨酸甲酯 盐酸盐, ≥98% (TLC), powder
Sigma-Aldrich
抗-GAPDH抗体,小鼠单克隆 小鼠抗, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
NG,NG-二甲基精氨酸 二盐酸盐
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)