• 主页
  • 查找结果
  • Direct counting of exosomes in a cell culture medium using neither isolation nor preconcentration.
Direct counting of exosomes in a cell culture medium using neither isolation nor preconcentration.
Analytica chimica acta (2020-05-23)
Tatsuya Fujii, Takashi Kaneta
摘要
Exosomes are expected to be biomarkers of cancer since they contain information about the cells that excrete them. In this study we developed a method to count the exosomes secreted from cancer cells in a culture medium without the need for isolation and/or preconcentration. This detection system consists of a square capillary on which a laser beam is focused in a sheet shape via the use of two cylindrical lenses. A fluorescently labeled anti-CD63 antibody is used to mark the exosomes that are then flowed into the square capillary. In this study, individual exosomes were observed on a trajectory when passing through the laser beam sheet and were counted for 10 min at a constant flow velocity. The total analysis time was less than 1.5 h including the steps required to remove large particles and allow reaction with the antibody. The results for two samples prepared with and without the isolation of exosomes showed a loss of exosomes in the isolation step. We also determined the number of the exosomes secreted by the cells to a culture medium during cultivation. As expected, the total number of exosomes in a culture medium increased with an increase in the cultivation time, and the number of exosomes released every 12 h either remained constant or showed no more than a slight increase for as long as 72 h. It was unclear whether the number exosomes was dependent on the cell population at confluences of 10-60%.
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。