• 主页
  • 查找结果
  • Lipid-polymer hybrid nanoparticles carrying linezolid improve treatment of methicillin-resistant Staphylococcus aureus (MRSA) harbored inside bone cells and biofilms.
Lipid-polymer hybrid nanoparticles carrying linezolid improve treatment of methicillin-resistant Staphylococcus aureus (MRSA) harbored inside bone cells and biofilms.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2020-04-27)
Pengbo Guo, Bettina A Buttaro, Hui Yi Xue, Ngoc T Tran, Ho Lun Wong
摘要
Methicillin-resistant Staphylococcus aureus (MRSA) is the most prevalent pathogen causing osteomyelitis. The tendency of MRSA to evade standard antibiotic treatment by hiding inside bone cells and biofilms is a major cause of frequent osteomyelitis recurrence. In this study, we developed a lipid-polymer hybrid nanoparticle loading the antibiotic linezolid (LIN-LPN), and focused on evaluating if this new nanoantibiotic can achieve significant in vitro activities against these intracellular and biofilm-embedded MRSA. The optimal LIN-LPN formulation demonstrated both high linezolid payload (12.0% by weight of nanoparticles) and controlled release characteristics (gradually released the entrapped antibiotic in 120 h). Although it achieved lower activities against bacteria including USA300-0114, CDC-587, RP-62A in planktonic form, it was substantially superior against the intracellular MRSA reservoir inside osteoblast cells. The differences of intracellular activities between LIN-LPN and linezolid were 87.0-fold, 12.3-fold, and 12.6-fold in CFU/ml (p < 0.05 or < 0.01) at 2 µg/ml, 4 µg/ml, and 8 µg/ml linezolid concentrations, respectively. LIN-LPN also suppressed the MRSA biofilm growth to 35-60% of the values achieved with free linezolid (p < 0.05). These enhanced intracellular and anti-biofilm activities of LIN-LPN were likely contributed by the extensive accumulation of LIN-LPN inside the MRSA-infected osteoblasts and biofilms as revealed in the confocal microscope images. The study thus validates the feasibility of exploiting the good nanoparticle-host cell and nanoparticle-biofilm interactions for improving the antibiotic drug activities against the poorly accessible bacteria, and supports LIN-LPN as a new alternative therapy for preventing the recurrence of MRSA-mediated bone infections.
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。