• 主页
  • 查找结果
  • Phototransformation of perfluorooctane sulfonamide on natural clay minerals: A likely source of short chain perfluorocarboxylic acids.
Phototransformation of perfluorooctane sulfonamide on natural clay minerals: A likely source of short chain perfluorocarboxylic acids.
Journal of hazardous materials (2020-02-26)
Kun Lv, Wei Gao, Lingyi Meng, Qiao Xue, Haoting Tian, Yawei Wang, Guibin Jiang
摘要
In this study, a new phototransformation pathway for perfluorooctane sulfonamide (FOSA) and underlying degradation mechanisms are described. Phototransformation of FOSA in a natural clay mineral (montmorillonite) suspension was compared to that in an aqueous solution. Results showed that the presence of montmorillonite can significantly promote the transformation of FOSA to perfluocarboxylic acids (increasing rate). The phototransformation reaction was found to be initiated by the activation of adsorbed oxygen molecules on the surface of montmorillonite, which generate superoxide anion and hydroxyl radicals. Hydroxyl radicals can then attack FOSA adsorbed onto the surface of montmorillonite, promoting the transformation process. In this reaction, clay minerals played a dual role: providing hydroxyl radicals and concentrating FOSA on their surfaces. This helped to promote the contact and reaction between FOSA and hydroxyl radicals. This study provides the first evidence that heterogeneous oxidation of FOSA at the surface of natural clay minerals may act as an important source of perfluocarboxylic acids (PFCAs), especially short chain PFCAs (i.e. trifluoroacetic acid, TFA).
材料
货号
品牌
产品描述
Sigma-Aldrich
五氟丙酸, 97%
Sigma-Aldrich
全氟戊酸, 97%
Sigma-Aldrich
全氟庚酸, ≥97.0%
Sigma-Aldrich
十一氟己酸, ≥97.0% (T)
Sigma-Aldrich
全氟辛基磺酰胺, AldrichCPR