• 主页
  • 查找结果
  • Spontaneous collapse of palmitic acid films on an alkaline buffer containing calcium ions.
Spontaneous collapse of palmitic acid films on an alkaline buffer containing calcium ions.
Colloids and surfaces. B, Biointerfaces (2020-05-15)
Pin Zhang, Tiep Pham, Xin Zheng, Chang Liu, Paola Leon Plata, Petr Král, Wei Bu, Binhua Lin, Ying Liu
摘要
Understanding the interaction of ions with fatty acids is important to identify their roles in various bioprocesses and to build novel biomimetic systems. In this study, the molecular organization of palmitic acid (PA) films on alkaline buffer solutions (pH 7.4) with and without divalent Ca2+ was measured at a constant surface area using Langmuir troughs coupled with microscopy and X-ray interfacial techniques. Without Ca2+, PA molecules remained a monolayer organization; however, with Ca2+, formation of the inverted bilayers of PA-Ca2+ superstructures caused a spontaneous 2D to 3D transformation under no compression due to the strong interaction between PA and the divalent cation. Self-assembly of this highly-organized inverted bilayer superstructure involved a two-step process of nucleation and nuclei growth. During nucleation, densely packed PA and Ca2+ monolayer firstly corrugated and some of PA and Ca2+ molecules ejected out from the monolayer; the ejected molecules then reorganized and formed the inverted bilayer nuclei. Nucleation was followed by nuclei growth, during which PA and Ca2+ in the monolayer kept integrating into the inverted bilayer structure through molecule migration and PA rotation around Ca2+.
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。