  • Search Results
  • Bile acids aggravate nonalcoholic steatohepatitis and cardiovascular disease in SHRSP5/Dmcr rat model.
Bile acids aggravate nonalcoholic steatohepatitis and cardiovascular disease in SHRSP5/Dmcr rat model.
Experimental and molecular pathology (2020-04-05)
Shusei Yamamoto, Ikumi Sato, Natsuki Fukuhama, Natsumi Akiyama, Miku Sakai, Shota Kumazaki, Shang Ran, Satoshi Hirohata, Kazuya Kitamori, Yukio Yamori, Shogo Watanabe
ABSTRACT
Nonalcoholic steatohepatitis (NASH) is linked to an increased risk of cardiovascular disease, regardless of the risk factors in metabolic syndrome. However, the intermediary factors between NASH and cardiovascular disease are still unknown. A previous study revealed that serum and hepatic bile acid (BA) levels are increased in some NASH patients. We aimed to examine whether NASH and cardiovascular disease were aggravated by BA using an animal model. From 10 to 18 weeks of age, SHRSP5/Dmcr rats divided into 3 groups were fed 3 types of high-fat and high-cholesterol (HFC) diets which were changed in the cholic acid (CA) concentration (0%, 2%, or 4%). The nitro oxide synthase inhibition (L-NAME) was administered intraperitoneally from 16 to 18 weeks of age. The 4% CA groups showed the worst LV dysfunction and myocardial fibrosis, and demonstrated severe hepatic fibrosis and lipid depositions. In addition, a large amount of lipid accumulation was observed in the aortas of the 4% CA group, and NFκB and VCAM-1 gene expression levels were increased. These findings were not seen in the 0% CA group. In the SHRSP5/Dmcr rat model, NASH and cardiovascular disease were aggravated with increasing BAs concentrations in an HFC diet.
Sigma-Aldrich
油红O, certified by the Biological Stain Commission
Sigma-Aldrich
Nω-硝基-L-精氨酸甲酯 盐酸盐, ≥98% (TLC), powder