  • Search Results
  • Red Cabbage (Brassica oleracea) Ameliorates Diabetic Nephropathy in Rats.
Red Cabbage (Brassica oleracea) Ameliorates Diabetic Nephropathy in Rats.
Evidence-based complementary and alternative medicine : eCAM (2008-10-03)
Hazem A H Kataya, Alaaeldin A Hamza
ABSTRACT
The protective action against oxidative stress of red cabbage (Brassica oleracea) extract was investigated. Diabetes was induced in male Wistar rats using streptozotocin (60 mg/kg body weight). Throughout the experimental period (60 days), diabetic rats exhibited many symptoms including loss of body weight, hyperglycemia, polyuria, polydipsia, renal enlargement and renal dysfunction. Significant increase in malondialdehyde, a lipid peroxidation marker, was observed in diabetic kidney. This was accompanied by a significant increase in reduced glutathione and superoxide dismutase activity and a decrease in catalase activity and in the total antioxidant capacity of the kidneys. Daily oral ingestion (1 g/kg body weight) of B. oleracea extract for 60 days reversed the adverse effect of diabetes in rats. B. oleracea extract lowered blood glucose levels and restored renal function and body weight loss. In addition, B. oleracea extract attenuated the adverse effect of diabetes on malondialdehyde, glutathione and superoxide dismutase activity as well as catalase activity and total antioxidant capacity of diabetic kidneys. In conclusion, the antioxidant and antihyperglycemic properties of B. oleracea extract may offer a potential therapeutic source for the treatment of diabetes.
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, suitable for (for molecular biology), Non-acetylated
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, crystallized, ≥98.0% (GE)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, powder, BioXtra
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, low endotoxin, Cohn Analog, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, pH <5.0, powder
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, chromatographically purified, New Zealand origin, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, ≥96% (agarose gel electrophoresis)