Merck
Clinically relevant cell sources for TMJ disc engineering.
Journal of dental research (2008-05-27)
D E Johns, M E Wong, K A Athanasiou
ABSTRACT
Tissue-engineering of the temporomandibular joint (TMJ) disc aims to provide patients with TMJ disorders an option to replace diseased tissue with autologous, functional tissue. This study examined clinically relevant cell sources by comparing costal chondrocytes, dermal fibroblasts, a mixture of the two, and TMJ disc cells in a scaffoldless tissue-engineering approach. It was hypothesized that all constructs would produce matrix relevant to the TMJ disc, but the mixture constructs were expected to appear most like the TMJ disc constructs. Costal chondrocyte and mixture constructs were morphologically and biochemically superior to the TMJ disc and dermal fibroblast constructs, and their compressive properties were not significantly different. Costal chondrocyte constructs produced almost 40 times more collagen and 800 times more glycosaminoglycans than did TMJ constructs. This study demonstrates the ability of costal chondrocytes to produce extracellular matrix that may function in a TMJ disc replacement.
Sigma-Aldrich
L -抗坏血酸, 99%
Supelco
维生素C, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L -抗坏血酸, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L -抗坏血酸, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L -抗坏血酸, ACS reagent, ≥99%
Sigma-Aldrich
胰岛素 人, recombinant, expressed in yeast (proprietary host)
Sigma-Aldrich
木瓜蛋白酶 来源于木瓜乳液, lyophilized powder, ≥10 units/mg protein
Sigma-Aldrich
L -抗坏血酸, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L -抗坏血酸, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
弹性蛋白酶 来源于猪胰腺, Type I, ≥4.0 units/mg protein
Sigma-Aldrich
L -抗坏血酸, reagent grade, crystalline
Sigma-Aldrich
木瓜蛋白酶 来源于木瓜乳液, buffered aqueous suspension, 2× Crystallized, ≥16 units/mg protein
Sigma-Aldrich
弹性蛋白酶 来源于猪胰腺, lyophilized powder, suitable for cell culture
Supelco
L -抗坏血酸, analytical standard
Sigma-Aldrich
L -抗坏血酸, meets USP testing specifications
Sigma-Aldrich
人胰岛素, recombinant, expressed in yeast, γ-irradiated, suitable for cell culture
Sigma-Aldrich
胰岛素 人, meets USP testing specifications
Sigma-Aldrich
木瓜蛋白酶 来源于番木瓜, powder, ≥3 U/mg
Sigma-Aldrich
L -抗坏血酸, reagent grade
Sigma-Aldrich
木瓜蛋白酶 来源于番木瓜, solution, light brown, ≥10 U/mg protein (~25 mg/ml)
Sigma-Aldrich
弹性蛋白酶 来源于猪胰腺, Type IV, Protein 50-90 %, lyophilized powder, ≥4.0 units/mg protein (biuret)
Sigma-Aldrich
木瓜蛋白酶 来源于木瓜乳液, crude powder, 1.5-10 units/mg solid
Sigma-Aldrich
L -抗坏血酸, FCC, FG
Sigma-Aldrich
L -抗坏血酸, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
弹性蛋白酶 来源于人类白血球, lyophilized powder, ≥50 units/mg protein (Bradford)
Sigma-Aldrich
木瓜蛋白酶 来源于木瓜乳液, lyophilized powder, aseptically filled
Sigma-Aldrich
L -抗坏血酸, tested according to Ph. Eur.
Sigma-Aldrich
L -抗坏血酸, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
弹性蛋白酶 来源于猪胰腺, Type III, lyophilized powder, Protein 55-85 %, ≥4.0 units/mg protein
Sigma-Aldrich
胰岛素 人, ≥95% (HPLC), semisynthetic, powder