• 主页
  • 查找结果
  • Tightening the retinal glia limitans attenuates neuroinflammation after optic nerve injury.
Tightening the retinal glia limitans attenuates neuroinflammation after optic nerve injury.
Glia (2020-07-10)
Evy Lefevere, Manuel Salinas-Navarro, Lien Andries, Lut Noterdaeme, Isabelle Etienne, Elien Van Wonterghem, Stefan Vinckier, Benjamin M Davis, Tine Van Bergen, Inge Van Hove, Kiavash Movahedi, Roosmarijn E Vandenbroucke, Lieve Moons, Lies De Groef
摘要
Increasing evidence suggests that functional impairments at the level of the neurovascular unit (NVU) underlie many neurodegenerative and neuroinflammatory diseases. While being part of the NVU, astrocytes have been largely overlooked in this context and only recently, tightening of the glia limitans has been put forward as an important neuroprotective response to limit these injurious processes. In this study, using the retina as a central nervous system (CNS) model organ, we investigated the structure and function of the glia limitans, and reveal that the blood-retina barrier and glia limitans function as a coordinated double barrier to limit infiltration of leukocytes and immune molecules. We provide in vitro and in vivo evidence for a protective response at the NVU upon CNS injury, which evokes inflammation-induced glia limitans tightening. Matrix metalloproteinase-3 (MMP-3) was found to be a crucial regulator of this process, thereby revealing its beneficial and immunomodulatory role in the CNS. in vivo experiments in which MMP-3 activity was deleted via genetic and pharmacological approaches, combined with a comprehensive study of tight junction molecules, glial end feet markers, myeloid cell infiltration, cytokine expression and neurodegeneration, show that MMP-3 attenuates neuroinflammation and neurodegeneration by tightening the glia limitans, thereby pointing to a prominent role of MMP-3 in preserving the integrity of the NVU upon injury. Finally, we gathered promising evidence to suggest that IL1b, which is also regulated by MMP-3, is at least one of the molecular messengers that induces glia limitans tightening in the injured CNS.
材料
货号
品牌
产品描述
Sigma-Aldrich
波形蛋白单克隆抗体 小鼠抗, clone VIM-13.2, ascites fluid
Sigma-Aldrich
NNGH, ≥98% (HPLC)
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。