• 主页
  • 查找结果
  • Atomic Force Microscope nanolithography on chromosomes to generate single-cell genetic probes.
Atomic Force Microscope nanolithography on chromosomes to generate single-cell genetic probes.
Journal of nanobiotechnology (2011-06-29)
Sebastiano Di Bucchianico, Anna M Poma, Maria F Giardi, Luana Di Leandro, Francesco Valle, Fabio Biscarini, Dario Botti
摘要
Chromosomal dissection provides a direct advance for isolating DNA from cytogenetically recognizable region to generate genetic probes for fluorescence in situ hybridization, a technique that became very common in cyto and molecular genetics research and diagnostics. Several reports describing microdissection methods (glass needle or a laser beam) to obtain specific probes from metaphase chromosomes are available. Several limitations are imposed by the traditional methods of dissection as the need for a large number of chromosomes for the production of a probe. In addition, the conventional methods are not suitable for single chromosome analysis, because of the relatively big size of the microneedles. Consequently new dissection techniques are essential for advanced research on chromosomes at the nanoscale level. We report the use of Atomic Force Microscope (AFM) as a tool for nanomanipulation of single chromosomes to generate individual cell specific genetic probes. Besides new methods towards a better nanodissection, this work is focused on the combination of molecular and nanomanipulation techniques which enable both nanodissection and amplification of chromosomal and chromatidic DNA. Cross-sectional analysis of the dissected chromosomes reveals 20 nm and 40 nm deep cuts. Isolated single chromosomal regions can be directly amplified and labeled by the Degenerate Oligonucleotide-Primed Polymerase Chain Reaction (DOP-PCR) and subsequently hybridized to chromosomes and interphasic nuclei. Atomic force microscope can be easily used to visualize and to manipulate biological material with high resolution and accuracy. The fluorescence in situ hybridization (FISH) performed with the DOP-PCR products as test probes has been tested succesfully in avian microchromosomes and interphasic nuclei. Chromosome nanolithography, with a resolution beyond the resolution limit of light microscopy, could be useful to the construction of chromosome band libraries and to the molecular cytogenetic mapping related to the investigation of genetic diseases.
材料
货号
品牌
产品描述
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, ≥250 units/mg solid
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, lyophilized powder, ≥2,500 units/mg protein (E1%/280)
Sigma-Aldrich
秋水仙碱, ≥95% (HPLC), powder
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, ≥400 units/mg protein
Sigma-Aldrich
秋水仙碱, BioReagent, suitable for plant cell culture, ≥95% (HPLC)