• 主页
  • 查找结果
  • Endogenous Chemerin from PVAT Amplifies Electrical Field-Stimulated Arterial Contraction: Use of the Chemerin Knockout Rat.
Endogenous Chemerin from PVAT Amplifies Electrical Field-Stimulated Arterial Contraction: Use of the Chemerin Knockout Rat.
International journal of molecular sciences (2020-09-06)
Emma D Flood, Stephanie W Watts
摘要
Background: We previously reported that the adipokine chemerin, when added exogenously to the isolated rat mesenteric artery, amplified electrical field-stimulated (EFS) contraction. The Chemerin1 antagonist CCX832 alone inhibited EFS-induced contraction in tissues with but not without perivascular adipose tissue (PVAT). These data suggested indirectly that chemerin itself, presumably from the PVAT, facilitated EFS-induced contraction. We created the chemerin KO rat and now test the focused hypothesis that endogenous chemerin amplifies EFS-induced arterial contraction. Methods: The superior mesenteric artery +PVAT from global chemerin WT and KO female rats, with endothelium and sympathetic nerve intact, were mounted into isolated tissue baths for isometric and EFS-induced contraction. Results: CCX832 reduced EFS (2-20 Hz)-induced contraction in tissues from the WT but not KO rats. Consistent with this finding, the magnitude of EFS-induced contraction was lower in the tissues from the KO vs. WT rats, yet the maximum response to the adrenergic stimulus PE was not different among all tissues. Conclusion: These studies support that endogenous chemerin modifies sympathetic nerve-mediated contraction through Chemerin1, an important finding relative in understanding chemerin's role in control of blood pressure.
材料
货号
品牌
产品描述
Sigma-Aldrich
白蛋白 来源于鸡蛋白, lyophilized powder, ≥90% (agarose gel electrophoresis)