  • Search Results
  • The Gustatory Sensory G-Protein GNAT3 Suppresses Pancreatic Cancer Progression in Mice.
The Gustatory Sensory G-Protein GNAT3 Suppresses Pancreatic Cancer Progression in Mice.
Cellular and molecular gastroenterology and hepatology (2020-09-04)
Megan T Hoffman, Samantha B Kemp, Daniel J Salas-Escabillas, Yaqing Zhang, Nina G Steele, Stephanie The, Daniel Long, Simone Benitz, Wei Yan, Robert F Margolskee, Filip Bednar, Marina Pasca di Magliano, Hui-Ju Wen, Howard C Crawford
ABSTRACT
Pancreatic ductal adenocarcinoma (PDA) initiation and progression are accompanied by an immunosuppressive inflammatory response. Here, we evaluated the immunomodulatory role of chemosensory signaling in metaplastic tuft cells (MTCs) by analyzing the role of GNAT3, a gustatory pathway G-protein expressed by MTCs, during PDA progression. Gnat3-null (Gnat3-/-) mice were crossbred with animals harboring a Cre-inducible KrasLSL-G12D/+ allele with either Ptf1aCre/+ (KC) or tamoxifen-inducible Ptf1aCreERT/+ (KCERT) mice to drive oncogenic KRAS expression in the pancreas. Ex vivo organoid conditioned medium generated from KC and Gnat3-/-;KC acinar cells was analyzed for cytokine secretion. Experimental pancreatitis was induced in KCERT and Gnat3-/-;KCERT mice to accelerate tumorigenesis, followed by analysis using mass cytometry and single-cell RNA sequencing. To study PDA progression, KC and Gnat3-/-;KC mice were aged to morbidity or 52 weeks. Ablation of Gnat3 in KC organoids increased release of tumor-promoting cytokines in conditioned media, including CXCL1 and CXCL2. Analysis of Gnat3-/-;KCERT pancreata found altered expression of immunomodulatory genes in Cxcr2 expressing myeloid-derived suppressor cells (MDSCs) and an increased number of granulocytic MDSCs, a subset of tumor promoting MDSCs. Importantly, expression levels of CXCL1 and CXCL2, known ligands for CXCR2, were also elevated in Gnat3-/-;KCERT pancreata. Consistent with the tumor-promoting role of MDSCs, aged Gnat3-/-;KC mice progressed more rapidly to metastatic carcinoma compared with KC controls. Compromised gustatory sensing, achieved by Gnat3 ablation, enhanced the CXCL1/2-CXCR2 axis to alter the MDSC population and promoted the progression of metastatic PDA.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
泰莫西芬, ≥99%
Sigma-Aldrich
视黄酸, ≥98% (HPLC), powder
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
氢化可的松, BioReagent, suitable for cell culture
Sigma-Aldrich
胰岛素 人, recombinant, expressed in yeast (proprietary host)
Sigma-Aldrich
hBFGF, FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
胶原酶 来源于溶组织梭菌, Type V, ≥1 FALGPA units/mg solid, >125 CDU/mg solid
Sigma-Aldrich
抗 α-淀粉酶 兔抗, fractionated antiserum, lyophilized powder