Merck
Ablation of Fat Cells in Adult Mice Induces Massive Bone Gain.
Cell metabolism (2020-10-08)
Wei Zou, Nidhi Rohatgi, Jonathan R Brestoff, Yongjia Li, Ruteja A Barve, Eric Tycksen, Yung Kim, Matthew J Silva, Steven L Teitelbaum
ABSTRACT
Adipocytes control bone mass, but the mechanism is unclear. To explore the effect of postnatal adipocyte elimination on bone cells, we mated mice expressing an inducible primate diphtheria toxin receptor (DTR) to those bearing adiponectin (ADQ)-Cre. DTR activation eliminates peripheral and marrow adipocytes in these DTRADQ mice. Within 4 days of DTR activation, the systemic bone mass of DTRADQ mice began to increase due to stimulated osteogenesis, with a 1,000% expansion by 10-14 days post-DTR treatment. This adipocyte ablation-mediated enhancement of skeletal mass reflected bone morphogenetic protein (BMP) receptor activation following the elimination of its inhibitors, associated with simultaneous epidermal growth factor (EGF) receptor signaling. DTRADQ-induced osteosclerosis is not due to ablation of peripheral adipocytes but likely reflects the elimination of marrow ADQ-expressing cells. Thus, anabolic drugs targeting BMP receptor inhibitors with short-term EGF receptor activation may be a means of profoundly increasing skeletal mass to prevent or reverse pathological bone loss.
Sigma-Aldrich
泰莫西芬, ≥99%
Sigma-Aldrich
地塞米松, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
L -抗坏血酸, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
胰岛素 人, recombinant, expressed in yeast (proprietary host)
Sigma-Aldrich
β-甘油磷酸盐 二钠盐 水合物, BioUltra, suitable for cell culture, suitable for plant cell culture, ≥99% (titration)
Sigma-Aldrich
单克隆抗 β-肌动蛋白抗体 小鼠抗, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
吲哚美辛, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
LDN193189 盐酸盐, ≥98% (HPLC)