• 主页
  • 查找结果
  • Analysis of Narrow-Leaf Lupin Proteins in Lupin-Enriched Pasta by Untargeted and Targeted Mass Spectrometry.
Analysis of Narrow-Leaf Lupin Proteins in Lupin-Enriched Pasta by Untargeted and Targeted Mass Spectrometry.
Foods (Basel, Switzerland) (2020-08-14)
Gilda Aiello, Yuchen Li, Giovanna Boschin, Marco Stanziale, Carmen Lammi, Anna Arnoldi
摘要
The supplementation of different food items with grain legumes and, in particular, with lupin has been demonstrated to provide useful health benefits, especially in the area of cardiovascular disease prevention. In this work, label free quantitative untargeted and targeted approaches based on liquid chromatography-electrospray ionization-tandem mass spectrometry (LC-ESI-MS/MS) for investigating the protein profile of three pasta samples containing different percentages of narrow-leaf lupin flour were carried out. The untargeted method permitted the identification of the main acidic globulins (α-conglutin, β-conglutin, and δ-conglutin) and the comparison of their profile with raw lupin flour. The targeted method, based on High-performance liquid chromatography electrospray ionization tandem mass spectrometry HPLC-Chip-Multiple Reaction Monitoring (MRM) mode, allowed the quantification of γ-conglutin, the main hypoglycemic component of lupin protein: its concentration was around 2.25 mg/g in sample A, 2.16 mg/g in sample D, and 0.57 mg/g in sample F.
材料
货号
品牌
产品描述
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, lyophilized powder, ≥2,500 units/mg protein (E1%/280)