Merck
  • 主页
  • 查找结果
  • Adenoviral vaccines promote protective tissue-resident memory T cell populations against cancer.
Adenoviral vaccines promote protective tissue-resident memory T cell populations against cancer.
Journal for immunotherapy of cancer (2020-12-10)
Esmé Ti van der Gracht, Mark Ja Schoonderwoerd, Suzanne van Duikeren, Ayse N Yilmaz, Felix M Behr, Julia M Colston, Lian N Lee, Hideo Yagita, Klaas Pjm van Gisbergen, Lukas Jac Hawinkels, Frits Koning, Paul Klenerman, Ramon Arens
摘要
Adenoviral vectors emerged as important platforms for cancer immunotherapy. Vaccination with adenoviral vectors is promising in this respect, however, their specific mechanisms of action are not fully understood. Here, we assessed the development and maintenance of vaccine-induced tumor-specific CD8+ T cells elicited upon immunization with adenoviral vectors. Adenoviral vaccine vectors encoding the full-length E7 protein from human papilloma virus (HPV) or the immunodominant epitope from E7 were generated, and mice were immunized intravenously with different quantities (107, 108 or 109 infectious units). The magnitude, kinetics and tumor protection capacity of the induced vaccine-specific T cell responses were evaluated. The adenoviral vaccines elicited inflationary E7-specific memory CD8+ T cell responses in a dose-dependent manner. The magnitude of these vaccine-specific CD8+ T cells in the circulation related to the development of E7-specific CD8+ tissue-resident memory T (TRM) cells, which were maintained for months in multiple tissues after vaccination. The vaccine-specific CD8+ T cell responses conferred long-term protection against HPV-induced carcinomas in the skin and liver, and this protection required the induction and accumulation of CD8+ TRM cells. Moreover, the formation of CD8+ TRM cells could be enhanced by temporal targeting CD80/CD86 costimulatory interactions via CTLA-4 blockade early after immunization. Together, these data show that adenoviral vector-induced CD8+ T cell inflation promotes protective TRM cell populations, and this can be enhanced by targeting CTLA-4.
材料
货号
品牌
产品描述
Sigma-Aldrich
DL-二硫苏糖醇, ≥98% (HPLC), ≥99.0% (titration)
Roche
Liberase TL研究级, lyophilized, suitable for tissue processing, optimum pH 7.4
Sigma-Aldrich
胶原酶 来源于溶组织梭菌, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
脱氧核糖核酸酶 I 来源于牛胰腺, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
分散酶®II, protease
Sigma-Aldrich
胶原酶 来源于溶组织梭菌, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid
Sigma-Aldrich
FTY720, ≥98% (HPLC)