• 主页
  • 查找结果
  • Formation of a novel coating material containing lutein and zeaxanthin via a Maillard reaction between bovine serum albumin and fucoidan.
Formation of a novel coating material containing lutein and zeaxanthin via a Maillard reaction between bovine serum albumin and fucoidan.
Food chemistry (2020-11-10)
Sunbin Kim, Weon-Sun Shin
摘要
The effective delivery of bioactive compounds has recently been receiving attention. In this study, a conjugate with BSA and fucoidan synthesized via the Maillard reaction was confirmed through electrophoresis, the o-phthalaldehyde assay, and through changes in absorbance. Two moles of fucoidan were glycated with one mole of BSA at 60 °C and 79% relative humidity for 4 days. The droplet coated with B-F conjugate remained stable during storage at 4 and 25 °C and slightly increased only at 55 °C however, the droplet coated with intact BSA and B/F mixture significantly increased. L/Z were degraded about 82, 79, and 36% for 4, 25, and 55 °C, respectively, regardless of the type of emulsifier. Although the conjugates could not prevent the degradation of lutein and zeaxanthin during storage, they improved the stability of the emulsion and showed 4.20-fold and 1.32-fold higher bioaccessibility than intact BSA and B/F mixtures, respectively.
材料
货号
品牌
产品描述
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, lyophilized powder, ≥2,500 units/mg protein (E1%/280)
Sigma-Aldrich
脂肪酶 来源于猪胰腺, Type II, 100-650 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
胆汁提取物 猪
Sigma-Aldrich
酚妥拉明 盐酸盐, ≥98% (TLC), powder