  • Search Results
  • Mechanism-based inhibitors of SIRT2: structure-activity relationship, X-ray structures, target engagement, regulation of α-tubulin acetylation and inhibition of breast cancer cell migration.
Mechanism-based inhibitors of SIRT2: structure-activity relationship, X-ray structures, target engagement, regulation of α-tubulin acetylation and inhibition of breast cancer cell migration.
RSC chemical biology (2021-08-31)
Alexander L Nielsen, Nima Rajabi, Norio Kudo, Kathrine Lundø, Carlos Moreno-Yruela, Michael Bæk, Martin Fontenas, Alessia Lucidi, Andreas S Madsen, Minoru Yoshida, Christian A Olsen
ABSTRACT
Sirtuin 2 (SIRT2) is a protein deacylase enzyme that removes acetyl groups and longer chain acyl groups from post-translationally modified lysine residues. It affects diverse biological functions in the cell and has been considered a drug target in relation to both neurodegenerative diseases and cancer. Therefore, access to well-characterized and robust tool compounds is essential for the continued investigation of the complex functions of this enzyme. Here, we report a collection of chemical probes that are potent, selective, stable in serum, water-soluble, and inhibit SIRT2-mediated deacetylation and demyristoylation in cells. Compared to the current landscape of SIRT2 inhibitors, this is a unique ensemble of features built into a single compound. We expect the developed chemotypes to find broad application in the interrogation of SIRT2 functions in both healthy and diseased cells, and to provide a foundation for the development of future therapeutics.
Product Description
Roche
不含EDTA的cOmplete蛋白酶抑制剂混合物, Tablets provided in glass vials
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
人血清, from human male AB plasma, USA origin, sterile-filtered
Sigma-Aldrich
胰蛋白酶 来源于牛胰腺, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
β-磷酸烟酰胺腺嘌呤二核苷酸 水合物