Merck
IFN-gamma reverses IL-2- and IL-4-mediated T-cell steroid resistance.
American journal of respiratory cell and molecular biology (2008-09-09)
Elena Goleva, Ling-Bo Li, Donald Y M Leung
ABSTRACT
Corticosteroids are the most common therapeutic approach for control of tissue inflammation. Combination IL-2/IL-4 is known to induce T-cell steroid resistance. This can be reversed with IFN-gamma; however, the mechanism by which this occurs is unknown. In the current study, we found that treatment of peripheral blood mononuclear cells with combination IL-2/IL-4 for 48 hours, but not with IL-2 or IL-4 alone, abrogated dexamethasone (DEX)-induced glucocorticoid receptor (GCR)-alpha nuclear translocation in both CD4(+) and CD8(+) T cells. The presence of IL-4 significantly down-regulated IFN-gamma production by IL-2-stimulated cells. Importantly, addition of IFN-gamma to the IL-2/IL-4 combination restored GCRalpha nuclear translocation in response to DEX. Furthermore, DEX-induced mitogen-activated protein kinase (MAPK) phosphatase-1 induction, used as a readout for corticosteroid-induced transactivation, was significantly greater (P < 0.05) in media and IL-2/IL-4/IFN-gamma-treated conditions compared with IL-2/IL-4-treated cells. The combination of IL-2/IL-4 induced p38 MAPK activation in CD3(+) cells (30.5 +/- 5.7% cells expressed phospho-p38 MAPK versus no phospho-p38 MAPK expression after media treatment). The presence of the p38 MAPK inhibitor, SB203580, or IFN-gamma inhibited p38 MAPK phosphorylation and enhanced GCRalpha nuclear translocation in response to DEX. These data indicate that combination IL-2/IL-4 inhibits GCRalpha nuclear translocation in human T cells, and this effect is reversed by IFN-gamma via inhibition of p38 MAPK activation.
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
地塞米松, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
地塞米松, ≥98% (HPLC), powder
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, suitable for (for molecular biology), Non-acetylated
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
皂素, for molecular biology, used as non-ionic surfactant
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
皂素, used as non-ionic surfactant
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, powder, BioXtra
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, crystallized, ≥98.0% (GE)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, pH <5.0, powder
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, low endotoxin, Cohn Analog, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%