  • Search Results
  • Adiponectin-coated nanoparticles for enhanced imaging of atherosclerotic plaques.
Adiponectin-coated nanoparticles for enhanced imaging of atherosclerotic plaques.
International journal of nanomedicine (2011-07-15)
Gunter Almer, Karin Wernig, Matthias Saba-Lepek, Samih Haj-Yahya, Johannes Rattenberger, Julian Wagner, Kerstin Gradauer, Daniela Frascione, Georg Pabst, Gerd Leitinger, Harald Mangge, Andreas Zimmer, Ruth Prassl
ABSTRACT
Atherosclerosis is a leading cause of mortality in the Western world, and plaque diagnosis is still a challenge in cardiovascular medicine. The main focus of this study was to make atherosclerotic plaques visible using targeted nanoparticles for improved imaging. Today various biomarkers are known to be involved in the pathophysiologic scenario of atherosclerotic plaques. One promising new candidate is the globular domain of the adipocytokine adiponectin (gAd), which was used as a targeting sequence in this study. gAd was coupled to two different types of nanoparticles, namely protamine-oligonucleotide nanoparticles, known as proticles, and sterically stabilized liposomes. Both gAd-targeted nanoparticles were investigated for their potency to characterize critical scenarios within early and advanced atherosclerotic plaque lesions using an atherosclerotic mouse model. Aortic tissue from wild type and apolipoprotein E-deficient mice, both fed a high-fat diet, were stained with either fluorescent-labeled gAd or gAd-coupled nanoparticles. Ex vivo imaging was performed using confocal laser scanning microscopy. gAd-targeted sterically stabilized liposomes generated a strong signal by accumulating at the surface of atherosclerotic plaques, while gAd-targeted proticles became internalized and showed more spotted plaque staining. Our results offer a promising perspective for enhanced in vivo imaging using gAd-targeted nanoparticles. By means of nanoparticles, a higher payload of signal emitting molecules could be transported to atherosclerotic plaques. Additionally, the opportunity is opened up to visualize different regions in the plaque scenario, depending on the nature of the nanoparticle used.
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, suitable for (for molecular biology), Non-acetylated
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
1,4-二叠氮双环[2.2.2]辛烷, ReagentPlus®, ≥99%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, crystallized, ≥98.0% (GE)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, powder, BioXtra
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, low endotoxin, Cohn Analog, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, pH <5.0, powder
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, chromatographically purified, New Zealand origin, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)