  • Search Results
  • Peroxisome proliferator-activated receptor-gamma agonists suppress tissue factor overexpression in rat balloon injury model with paclitaxel infusion.
Peroxisome proliferator-activated receptor-gamma agonists suppress tissue factor overexpression in rat balloon injury model with paclitaxel infusion.
PloS one (2011-12-06)
Jun-Bean Park, Baek-Kyung Kim, Yoo-Wook Kwon, Dominik N Muller, Hyun-Chae Lee, Seock-Won Youn, Young-Eun Choi, Sae-Won Lee, Han-Mo Yang, Hyun-Jai Cho, Kyung Woo Park, Hyo-Soo Kim
ABSTRACT
The role and underlying mechanisms of rosiglitazone, a peroxisome proliferator-activated receptor-gamma (PPAR-γ) agonist, on myocardial infarction are poorly understood. We investigated the effects of this PPAR-γ agonist on the expression of tissue factor (TF), a primary molecule for thrombosis, and elucidated its underlying mechanisms. The PPAR-γ agonist inhibited TF expression in response to TNF-α in human umbilical vein endothelial cells, human monocytic leukemia cell line, and human umbilical arterial smooth muscle cells. The overexpression of TF was mediated by increased phosphorylation of mitogen-activated protein kinase (MAPK), which was blocked by the PPAR-γ agonist. The effective MAPK differed depending on each cell type. Luciferase and ChIP assays showed that transcription factor, activator protein-1 (AP-1), was a pivotal target of the PPAR-γ agonist to lower TF transcription. Intriguingly, two main drugs for drug-eluting stent, paclitaxel or rapamycin, significantly exaggerated thrombin-induced TF expression, which was also effectively blocked by the PPAR-γ agonist in all cell types. This PPAR-γ agonist did not impair TF pathway inhibitor (TFPI) in three cell types. In rat balloon injury model (Sprague-Dawley rats, n = 10/group) with continuous paclitaxel infusion, the PPAR-γ agonist attenuated TF expression by 70±5% (n = 4; P<0.0001) in injured vasculature. Taken together, rosiglitazone reduced TF expression in three critical cell types involved in vascular thrombus formation via MAPK and AP-1 inhibitions. Also, this PPAR-γ agonist reversed the paclitaxel-induced aggravation of TF expression, which suggests a possibility that the benefits might outweigh its risks in a group of patients with paclitaxel-eluting stent implanted.
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, suitable for (for molecular biology), Non-acetylated
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, crystallized, ≥98.0% (GE)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, powder, BioXtra
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, low endotoxin, Cohn Analog, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, pH <5.0, powder
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, chromatographically purified, New Zealand origin, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, ≥98% (agarose gel electrophoresis)