• 主页
  • 查找结果
  • Immunohistochemical localization of calpains and calpastatin in the rabbit eye.
Immunohistochemical localization of calpains and calpastatin in the rabbit eye.
Brain research (1993-05-21)
H Persson, S Kawashima, J O Karlsson
摘要
The localization of the two Ca-activated extralysosomal proteases m-calpain and mu-calpain in the eye of the adult rabbit was examined by immunohistochemistry, using poly- and monoclonal antibodies against the corresponding rabbit antigens. Immunoreactivity against the two forms of calpains was observed in the epithelial cells on the external and internal surface of the cornea as well as in the epithelial cells covering the iris and ciliary body. The sclera and choroid layers showed a relatively weak immunoreactivity. Using anti m-calpain antibodies, the pigment epithelium in the retina was heavily labelled as well as the outer and inner plexiform layers. The other and inner borders of the Müller cells were clearly labelled. The outer segments of the receptor cells showed a strong immunoreactivity for both mu-calpain and m-calpain. Labelling was also observed in the retinal ganglion cells and in the nerve fiber layer. The immunohistochemical localization of calpastatin, an endogenous inhibitor of both m- and mu-calpain was also examined. A high level of calpastatin immunoreactivity was observed in the outer segments of the receptor cells. The results may be compatible with a role for calpains, especially m-calpain, in the secretory/phagocytic process and as modulators of the cytoskeleton in cell processes.
材料
货号
品牌
产品描述
Sigma-Aldrich
Monoclonal Anti-m-Calpain (Domain III/IV) antibody produced in mouse, clone 107-82, ascites fluid
Sigma-Aldrich
单克隆抗-钙抑素 小鼠抗, clone 1F7E3D10, ascites fluid, buffered aqueous solution
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。