  • Search Results
  • In vivo stable tumor-specific painting in various colors using dehalogenase-based protein-tag fluorescent ligands.
In vivo stable tumor-specific painting in various colors using dehalogenase-based protein-tag fluorescent ligands.
Bioconjugate chemistry (2009-06-12)
Nobuyuki Kosaka, Mikako Ogawa, Peter L Choyke, Natasha Karassina, Cesear Corona, Mark McDougall, David T Lynch, Clifford C Hoyt, Richard M Levenson, Georgyi V Los, Hisataka Kobayashi
ABSTRACT
In vivo fluorescence cancer imaging is an important tool in understanding tumor growth and therapeutic monitoring and can be performed either with endogenously produced fluorescent proteins or with exogenously introduced fluorescent probes bound to targeting molecules. However, endogenous fluorescence proteins cannot be altered after transfection, thus requiring rederivation of cell lines for each desired color, while exogenously targeted fluorescence probes are limited by the heterogeneous expression of naturally occurring cellular targets. In this study, we adapted the dehalogenase-based protein-Tag (HaloTag) system to in vivo cancer imaging, by introducing highly expressed HaloTag receptors (HaloTagR) in cancer cells coupled with a range of externally injected fluorophore-conjugated dehalogenase-reactive reactive linkers. Tumor nodules arising from a single transfected cell line were stably labeled with fluorescence varying in emission spectra from green to near-infrared. After establishing and validating a SHIN3 cell line stably transfected with HaloTagR (HaloTagR-SHIN3), in vivo spectral fluorescence imaging studies were performed in live animals using a peritoneal dissemination model. The tumor nodules arising from HaloTagR-SHIN3 could be successfully labeled by four different fluorophore-conjugated HaloTag-ligands each emitting light at different wavelengths. These fluorophores could be alternated on serial imaging sessions permitting assessment of interval growth. Fluorescence was retained in histological specimens after fixation. Thus, this tagging system proves versatile both for in vivo and in vitro imaging without requiring modification of the underlying cell line. Thus, this strategy can overcome some of the limitations associated with the use of endogenous fluorescent proteins and exogenous targeted optical agents in current use.
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, suitable for (for molecular biology), Non-acetylated
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, crystallized, ≥98.0% (GE)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, powder, BioXtra
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, low endotoxin, Cohn Analog, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, pH <5.0, powder
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, chromatographically purified, New Zealand origin, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
牛血清白蛋白 来源于牛血清, lyophilized powder, ≥98% (agarose gel electrophoresis)