• 主页
  • 查找结果
  • Purification and identification of antioxidant peptides from walnut (Juglans regia L.) protein hydrolysates.
Purification and identification of antioxidant peptides from walnut (Juglans regia L.) protein hydrolysates.
Peptides (2012-10-02)
Ning Chen, Hongmei Yang, Yi Sun, Jun Niu, Shuying Liu
摘要
Walnut proteins were hydrolyzed separately using three different proteases to obtain antioxidant peptides. The antioxidant activities of the hydrolysates were measured using 1,1-diphenyl-2-picryl hydrazyl (DPPH) assay. Among hydrolysates, pepsin hydrolysate obtained by 3h exhibited the highest antioxidant activities, which could also quench the hydroxyl radical, chelate ferrous ion, exhibit reducing power and inhibit the lipid peroxidation. Then, 3-h pepsin hydrolysates were purified sequentially by ultrafiltration, gel filtration and RP-HPLC. The sequence of the peptide with the highest antioxidative activity was identified to be Ala-Asp-Ala-Phe (423.23 Da) using RP-HPLC-ESI-MS, which was identified for the first time from walnut protein hydrolysates. Last, the inhibition of the peptide on lipid peroxidation was similar with that of reduced glutathione (GSH). These results indicate that the protein hydrolysates and/or its isolated peptides may be effectively used as food additives.
材料
货号
品牌
产品描述
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, ≥250 units/mg solid
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, lyophilized powder, ≥2,500 units/mg protein (E1%/280)
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, ≥400 units/mg protein
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, slightly beige, ≥500 U/mg
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, slightly beige, 1200-2400 U/mg
Sigma-Aldrich
胃蛋白酶−琼脂糖 来源于猪胃粘膜, lyophilized powder, ≥30 units/mg dry solid