• 主页
  • 查找结果
  • Lung injury-dependent oxidative status and chymotrypsin-like activity of skeletal muscles in hamsters with experimental emphysema.
Lung injury-dependent oxidative status and chymotrypsin-like activity of skeletal muscles in hamsters with experimental emphysema.
BMC musculoskeletal disorders (2013-01-25)
Jair Tonon, Alessandra Lourenço Cecchini, Cláudia Roberta Brunnquell, Sara Santos Bernardes, Rubens Cecchini, Flávia Alessandra Guarnier
摘要
Peripheral skeletal muscle is altered in patients suffering from emphysema and chronic obstructive pulmonary disease (COPD). Oxidative stress have been demonstrated to participate on skeletal muscle loss of several states, including disuse atrophy, mechanical ventilation, and chronic diseases. No evidences have demonstrated the occurance in a severity manner. We evaluated body weight, muscle loss, oxidative stress, and chymotrypsin-like proteolytic activity in the gastrocnemius muscle of emphysemic hamsters. The experimental animals had 2 different severities of lung damage from experimental emphysema induced by 20 mg/mL (E20) and 40 mg/mL (E40) papain. The severity of emphysema increased significantly in E20 (60.52 ± 2.8, p < 0.05) and E40 (52.27 ± 4.7; crossed the alveolar intercepts) groups. As compared to the control group, there was a reduction on body (171.6 ± 15.9 g) and muscle weight (251.87 ± 24.87 mg) in the E20 group (157.5 ± 10.3 mg and 230.12 ± 23.52 mg, for body and muscle weight, respectively), which was accentuated in the E40 group (137.4 ± 7.2 g and 197.87 ± 10.49 mg, for body and muscle weight, respectively). Additionally, the thiobarbituric acid reactive substances (TBARS), tert-butyl hydroperoxide-initiated chemiluminescence (CL), carbonylated proteins, and chymotrypsin-like proteolytic activity were elevated in the E40 group as compared to the E20 group (p < 0.05 for all comparisons). The severity of emphysema significantly correlated with the progressive increase in CL (r = -0.95), TBARS (r = -0.98), carbonyl proteins (r = -0.99), and chymotrypsin-like proteolytic activity (r = -0.90). Furthermore, augmentation of proteolytic activity correlated significantly with CL (r = 0.97), TBARS (r = 0.96), and carbonyl proteins (r = 0.91). Taken together, the results of the present study suggest that muscle atrophy observed in this model of emphysema is mediated by increased muscle chymotrypsin-like activity, with possible involvement of oxidative stress in a severity-dependent manner.
材料
货号
品牌
产品描述
Sigma-Aldrich
Luperox® TBH70X, 丁基过氧化氢 溶液, 70 wt. % in H2O
Sigma-Aldrich
叔丁基过氧化氢 溶液, 5.0-6.0 M in decane
Sigma-Aldrich
α-糜蛋白酶 来源于牛胰腺, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
木瓜蛋白酶 来源于番木瓜, powder, ≥3 U/mg
Sigma-Aldrich
叔丁基过氧化氢 溶液, packed in FEP bottles, ~5.5 M in decane (over molecular sieve 4Å)
Sigma-Aldrich
木瓜蛋白酶 来源于木瓜乳液, lyophilized powder, aseptically filled
Sigma-Aldrich
木瓜蛋白酶 来源于番木瓜, solution, light brown, ≥10 U/mg protein (~25 mg/ml)
Sigma-Aldrich
木瓜蛋白酶 来源于木瓜乳液, crude powder, 1.5-10 units/mg solid
Sigma-Aldrich
α-糜蛋白酶 来源于牛胰腺, (TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
α-糜蛋白酶 来源于牛胰腺, Type I-S, essentially salt-free, lyophilized powder
Sigma-Aldrich
α-糜蛋白酶 来源于牛胰腺, suitable for protein sequencing, salt-free, lyophilized powder
Sigma-Aldrich
α-糜蛋白酶 来源于牛胰腺, ≥40 units/mg protein, vial of 5 mg
Sigma-Aldrich
叔丁基过氧化氢 溶液, 5.0-6.0 M in nonane
Sigma-Aldrich
α-糜蛋白酶 来源于人类胰腺, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin−Agarose from bovine pancreas, lyophilized powder, 2,000-3,500 units/g agarose (One ml gel will yield 65-120 units)