• 主页
  • 查找结果
  • Measurement of local strains in intervertebral disc anulus fibrosus tissue under dynamic shear: contributions of matrix fiber orientation and elastin content.
Measurement of local strains in intervertebral disc anulus fibrosus tissue under dynamic shear: contributions of matrix fiber orientation and elastin content.
Journal of biomechanics (2009-08-12)
Arthur J Michalek, Mark R Buckley, Lawrence J Bonassar, Itai Cohen, James C Iatridis
摘要
Shear strain has been implicated as an initiator of intervertebral disc anulus failure, however a clear, multi-scale picture of how shear strain affects the tissue microstructure has been lacking. The purposes of this study were to measure microscale deformations in anulus tissue under dynamic shear in two orientations, and to determine the role of elastin in regulating these deformations. Bovine AF tissue was simultaneously shear loaded and imaged using confocal microscopy following either a buffer or elastase treatment. Digital image analysis was used to track through time local shear strains in specimens sheared transversely, and stretch and rotation of collagen fiber bundles in specimens sheared circumferentially. The results of this study suggest that sliding does not occur between AF plies under shear, and that interlamellar connections are governed by collagen and fibrilin rather than elastin. The transverse shear modulus was found to be approximately 1.6 times as high in plies the direction of the collagen fibers as in plies across them. Under physiological levels of in-plane shear, fiber bundles stretched and re-oriented linearly. Elastin was found to primarily stiffen plies transversely. We conclude that alterations in the elastic fiber network, as found with IVD herniation and degeneration, can therefore be expected to significantly influence the AF response to shear making it more susceptible to micro failure under bending or torsion loading.
材料
货号
品牌
产品描述
Sigma-Aldrich
弹性蛋白酶 来源于猪胰腺, Type I, ≥4.0 units/mg protein
Sigma-Aldrich
弹性蛋白酶 来源于猪胰腺, lyophilized powder, suitable for cell culture
Sigma-Aldrich
弹性蛋白酶 来源于猪胰腺, Type IV, Protein 50-90 %, lyophilized powder, ≥4.0 units/mg protein (biuret)
Sigma-Aldrich
弹性蛋白酶 来源于猪胰腺, Type III, lyophilized powder, Protein 55-85 %, ≥4.0 units/mg protein