Pepsinogen D. A fourth proteolytic zymogen from pig gastric mucosa.
The Biochemical journal (1967-09-01)
D Lee, A P Ryle
摘要
Methods are described for the isolation and purification of pepsinogen D, a minor zymogen occurring to the extent of about 5% of the potential proteolytic activity in neutral extracts of the pig gastric mucosa. The physical and chemical properties of this zymogen indicate that it is very similar to, if not identical with, dephosphopepsinogen.
材料
货号
品牌
产品描述
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, ≥250 units/mg solid
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, lyophilized powder, ≥2,500 units/mg protein (E1%/280)
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, ≥400 units/mg protein
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, slightly beige, ≥500 U/mg
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, slightly beige, 1200-2400 U/mg
Sigma-Aldrich
胃蛋白酶−琼脂糖 来源于猪胃粘膜, lyophilized powder, ≥30 units/mg dry solid