Merck
Chemical constituents of the stems of Ephedra sinica.
Journal of Asian natural products research (2009-02-17)
Wei Zhao, An-Jun Deng, Guan-Hua Du, Jin-Lan Zhang, Zhi-Hong Li, Hai-Lin Qin
ABSTRACT
A new naphthalene derivative, 1-methyl-2,3-methylenedioxy-6-naphthalenecarboxylic acid methyl ester (1), and a new alkaloid, ( +/- )-1-phenyl-2-imido-1-propanol (2), together with the four known compounds, ephedrine, pseudoephedrine, N-methylephedrine, and 6-methoxykynurenic acid, have been isolated from the stems of Ephedra sinica, a famous traditional Chinese herbal medicine. The structures of 1 and 2 were determined by spectroscopic methods, including 1D- and 2D-NMR experiments as well as HR-EI-MS analysis.
Sigma-Aldrich
(1R,2S)-(-)-N-甲基麻黄碱, 99%
Supelco
甲基麻黄碱标准液 溶液, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®