• 主页
  • 查找结果
  • Association of adenovirus early-region 1A proteins with cellular polypeptides.
Association of adenovirus early-region 1A proteins with cellular polypeptides.
Molecular and cellular biology (1986-05-01)
E Harlow, P Whyte, B R Franza, C Schley
摘要
Extracts from adenovirus-transformed human 293 cells were immunoprecipitated with monoclonal antibodies specific for the early-region 1A (E1A) proteins. In addition to the E1A polypeptides, these antibodies precipitated a series of proteins with relative molecular weights of 28,000, 40,000, 50,000, 60,000, 80,000, 90,000, 110,000, 130,000, and 300,000. The two most abundant of these polypeptides are the 110,000-molecular-weight protein (110K protein) and 300K protein. Three experimental approaches have suggested that the 110K and 300K polypeptides are precipitated because they form stable complexes with the E1A proteins. The 110K and 300K polypeptides do not share epitopes with the E1A proteins, they copurify with a subset of the E1A proteins, and they bind to the E1A proteins following mixing in vitro. The 110K and 300K polypeptides are not adenoviral proteins, but are encoded by cellular DNA. Both the 12S and the 13S E1A proteins bind to the 110K and 300K species, and these complexes are found in adenovirus-transformed and -infected cells.
材料
货号
品牌
产品描述
Sigma-Aldrich
Monoclonal Anti-Adenovirus 2 E1A (Adenovirus 2 early region 1) antibody produced in mouse, 1.0 mg/mL, clone M73, purified immunoglobulin, buffered aqueous solution
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。