A new method for the preparation of monoporous hollow microspheres.
Langmuir : the ACS journal of surfaces and colloids (2010-01-26)
Ming-Wei Chang, Eleanor Stride, Mohan Edirisinghe
摘要
The feasibility of producing a hollow microsphere with a single hole in its shell by coaxial electrohydrodynamic atomization (CEHDA) is demonstrated. Polymethylsilsesquioxane (PMSQ) was used as a model shell material encapsulating a core of a volatile liquid, perfluorohexane (PFH), which was subsequently evaporated to produce the hollow microspheres. The diameters of the microspheres and of the single surface pore were controlled by varying the flow rate of the components, the concentration of the PMSQ solution, and the applied voltage in the CEHDA process. The particles were characterized by scanning electron microscopy, and the ranges obtained were 275-860 nm for the microsphere diameter and 35-135 nm for the pore size. The process overcomes several of the key problems associated with existing methods of monoporous microsphere formation including removing the need for elevated temperatures, multiple processing steps, and the use of surfactants and other additives.
材料
货号
品牌
产品描述
Sigma-Aldrich
十四氟己烷, 99%
Sigma-Aldrich
十四氟己烷, A mixture of perfluorinated hexanes, 95%