  • Search Results
  • Comparison of contents of five ephedrine alkaloids in three official origins of Ephedra Herb in China by high-performance liquid chromatography.
Comparison of contents of five ephedrine alkaloids in three official origins of Ephedra Herb in China by high-performance liquid chromatography.
Journal of natural medicines (2011-04-06)
Hao Hong, Hu-Biao Chen, Dong-Hui Yang, Ming-Ying Shang, Xuan Wang, Shao-Qing Cai, Masayuki Mikage
ABSTRACT
In the present paper, a fast and economical HPLC method [on a Phenomenex Polar-RP column with a solution of (phosphoric acid:triethylamine:dibutylamine:water = 0.40:0.1:0.2:499.3) and methanol] is developed, and applied for the determination of norephedrine, norpseudoephedrine, ephedrine (E), pseudoephedrine (PE) and methylephedrine (ME) in 64 samples of three species from main habitats in China. Quantitation data showed that total alkaloid content in Ephedra equisetina Bge. (2.708 ± 0.642%) is higher than that in E. sinica Stapf. (1.365 ± 0.624%) and E. intermedia Schrenk et C. A. Mey. (1.537 ± 0.746%), but the range of total alkaloid content in each species is so wide that the ranges of the three species greatly overlap. The contents of E, PE and ME are different among the three species. The ratio E/total alkaloid content and ratio E/PE as well as E and ME contents can be used for identification of E. sinica, E. intermedia and E. equisetina from one another.
Sigma-Aldrich
(1S,2R)-(+)-去甲麻黄素, 98%
Supelco
(±)-盐酸去甲伪麻黄碱标准液 盐酸盐 溶液, 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(1R,2S)-(-)-N-甲基麻黄碱, 99%
Sigma-Aldrich
DL-去甲麻黄素 盐酸盐, ≥99%
Supelco
(±)盐酸苯丙醇胺标准液 盐酸盐 溶液, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(1S,2S)-(+)-去甲伪麻黄碱, ≥98.0% (NT)
Supelco
甲基麻黄碱标准液 溶液, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Cathine hydrochloride