  • Search Results
  • [Determination of ephedrine hydrochloride, pseudoephedrine hydrochloride and methylephedrine hydrochloride in maxingshigan decoction by CE].
[Determination of ephedrine hydrochloride, pseudoephedrine hydrochloride and methylephedrine hydrochloride in maxingshigan decoction by CE].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2011-08-04)
Li-ping Yu, Xiao-ke Wang, Jia-bo Luo
ABSTRACT
To establish the method for determination of ephedrine hydrochloride, pseudoephedrine hydrochloride and methylephedrine hydrochloride in maxingshigan decoction by capillary electrophoresis. The separation was performed on a fused silica capillary of 60 cm x 55 microcrpm ID (52 cm of effective length). 60 mmol/L NaB4O7 + 10% (V/V) CH3OH (pH 9.0) was selected as the running buffer. The separation voltage was 12 kV. The samples was injected by gravity (10 s, 15 cm). The detection wavelength was 210 nm and berberine hydrochloride was the internal standard. The linear range of determination for ephedrine hydrochloride, pseudoephedrine hydrochloride and methylephedrine hydrochloride were 20.0-160.0 microg/mL (r = 0.9999), 7.5-60.0 microg/mL (r = 0.9991) and 2.0-10.0 microg/mL (r = 0.9993). The average recoveries were 98.0%, 97.0% and 97.8%, the precisions of the method were 2.31%, 2.21% and 2.00% (n=6), respectively. The method is convenient, rapid and accurate for the quality control of maxingshigan decoction.
Sigma-Aldrich
(1R,2S)-(-)-N-甲基麻黄碱, 99%
Supelco
甲基麻黄碱标准液 溶液, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®