• 主页
  • 查找结果
  • Enhanced stability of Cu-BTC MOF via perfluorohexane plasma-enhanced chemical vapor deposition.
Enhanced stability of Cu-BTC MOF via perfluorohexane plasma-enhanced chemical vapor deposition.
Journal of the American Chemical Society (2012-01-14)
Jared B Decoste, Gregory W Peterson, Martin W Smith, Corinne A Stone, Colin R Willis
摘要
Metal organic frameworks (MOFs) are a leading class of porous materials for a wide variety of applications, but many of them have been shown to be unstable toward water. Cu-BTC (1,3,5 benzenetricarboxylic acid, BTC) was treated with a plasma-enhanced chemical vapor deposition (PECVD) of perfluorohexane creating a hydrophobic form of Cu-BTC. It was found that the treated Cu-BTC could withstand high humidity and even submersion in water much better than unperturbed Cu-BTC. Through Monte Carlo simulations it was found that perfluorohexane sites itself in such a way within Cu-BTC as to prevent the formation of water clusters, hence preventing the decomposition of Cu-BTC by water. This PECVD of perfluorohexane could be exploited to widen the scope of practical applications of Cu-BTC and other MOFs.
材料
货号
品牌
产品描述
Sigma-Aldrich
苯均三酸, 95%
Sigma-Aldrich
十四氟己烷, 99%
Sigma-Aldrich
十四氟己烷, A mixture of perfluorinated hexanes, 95%