• 主页
  • 查找结果
  • Shear bond strength in zirconia veneered ceramics using two different surface treatments prior veneering.
Shear bond strength in zirconia veneered ceramics using two different surface treatments prior veneering.
Collegium antropologicum (2013-05-24)
Lana Bergman Gasparić, Zdravko Schauperl, Ketij Mehulić
摘要
Aim of the study was to assess the effect of different surface treatments on the shear bond strength (SBS) of the veneering ceramics to zirconia core. In a shear test the influence of grinding and sandblasting of the zirconia surface on bonding were assessed. Statistical analysis was performed using SPSS statistical package (version 17.0, SPSS Inc., Chicago, IL, USA) and Microsoft Office Excel 2003 (Microsoft, Seattle, WA, USA). There was a significant difference between the groups considering shear bond strength (SBS) values, i.e. ground and sandblasted samples had significantly higher SBS values than only ground samples (mean difference = -190.67; df = 10, t = -6.386, p < 0.001). The results of the present study indicate that ground and sandblasted cores are superior to ground cores, allowing significantly higher surface roughness and significantly higher shear bond strength between the core and the veneering material.
材料
货号
品牌
产品描述
Sigma-Aldrich
聚四氟乙烯 制备, 60 wt % dispersion in H2O
Sigma-Aldrich
氧化锆 (IV), powder, 5 μm, 99% trace metals basis
Sigma-Aldrich
聚四氟乙烯, powder (free-flowing), 1 μm particle size
Sigma-Aldrich
氧化锆 (IV), nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
聚四氟乙烯, powder, >40 μm particle size
Sigma-Aldrich
锆, powder, −100 mesh
Sigma-Aldrich
聚四氟乙烯, powder, 200 μm particle size
Sigma-Aldrich
聚四氟乙烯, beads
Sigma-Aldrich
聚四氟乙烯, powder, 35 μm particle size
Sigma-Aldrich
锆, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
聚四氟乙烯, powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
氧化锆(IV), nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O
Sigma-Aldrich
氧化锆 (IV), 99.99% trace metals basis (purity excludes ~2% HfO2)
Sigma-Aldrich
锆, sponge, ≥99% trace metals basis
Sigma-Aldrich
锆, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
聚四氟乙烯, powder, ≥350 μm particle size
Sigma-Aldrich
氧化锆(IV), nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O
Sigma-Aldrich
锆, powder, in water, -325 mesh, 99.5% trace metals basis (excluding Hf)