• 主页
  • 查找结果
  • The effect of oleic acid stabilizer on the surface properties of bimetallic PtNi catalysts.
The effect of oleic acid stabilizer on the surface properties of bimetallic PtNi catalysts.
Journal of nanoscience and nanotechnology (2013-08-02)
N H H Abu Bakar, M Abu Bakar, M M Bettahar, J Ismail, S Monteverdi
摘要
A detailed study on the surface properties of oleic acid-stabilized PtNi nanoparticles supported on silica is reported. The oleic acid-stabilized PtNi nanoparticles were synthesized using NaBH4 as the reducing agent at various temperatures and oleic acid concentrations, prior to incorporation onto the silica support. X-ray diffraction studies of the unsupported oleic acid-stabilized PtNi particles revealed that the PtNi existed as alloys. Upon incorporation onto silica support, surface properties of the catalysts were investigated using H2-temperature reduction (H2-TPR), H2-temperature desorption (H2-TPD) and H2-chemisorption techniques. It was found that for the bimetallic catalysts, no oxides or very little oxidation occurred. Furthermore, these catalysts exhibited both Pt and Ni active sites on its surface though the availability of Ni active sites was dominant. A comparison of the surface properties of these materials with those prepared without oleic acid in our previous work [N. H. H. Abu Bakar et al., J. Catal. 265, 63 (2009)] and how they affect the hydrogenation of benzene is also discussed.
材料
货号
品牌
产品描述
Sigma-Aldrich
油酸, ≥99% (GC)
Sigma-Aldrich
油酸, technical grade, 90%
Sigma-Aldrich
油酸, suitable for cell culture, BioReagent
Supelco
油酸, analytical standard
Sigma-Aldrich
Raney®-镍, W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst
Sigma-Aldrich
油酸, natural, FCC
Sigma-Aldrich
铂, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
镍, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
镍, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
铂, wire, diam. 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
铂, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
铂, gauze, 100 mesh, 99.9% trace metals basis
Sigma-Aldrich
铂, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
铂黑, black, powder, ≤20 μm, ≥99.95% trace metals basis
Sigma-Aldrich
铂, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
镍, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
铂, gauze, 52 mesh, 99.9% trace metals basis
Sigma-Aldrich
镍, foil, thickness 0.125 mm, ≥99.9%
Sigma-Aldrich
油酸, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Raney®-镍, W.R. Grace and Co. Raney® 4200, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-镍, W.R. Grace and Co. Raney® 2400, slurry, in H2O, active catalyst
Sigma-Aldrich
铂, powder, 99.995% trace metals basis
Sigma-Aldrich
镍, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
镍, powder, <1 μm, 99.8% trace metals basis
Sigma-Aldrich
铂, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
镍, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
铂, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
铂, wire, diam. 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
铂, wire, diam. 0.076 mm, ≥99.99% trace metals basis
Sigma-Aldrich
铂, powder, ≤10 μm, 99.9% trace metals basis