Deconjugation of glucuronides catalysed by UDPglucuronyltransferase.
Biochimica et biophysica acta (1986-09-26)
W H Peters, P L Jansen, H T Cuypers, R A de Abreu, H Nauta
摘要
Evidence was found for UDPglucuronyltransferase-catalysed deconjugation of p-nitrophenol-, 4-methylumbelliferone- and phenolphthalein-glucuronides. The evidence is based on the following observations: 1, deconjugation is UDP-dependent and the reactions show Michaels-Menten kinetics with respect to UDP and glucuronide saturability; 2, UDP-glucuronic acid was identified as reaction product; 3, all studies were done in the presence of a beta-glucuronidase inhibitor; 4, induction profiles, using 3-methylcholanthrene and phenobarbital as inducing agents, were identical for conjugation and deconjugation reactions. Optimal deconjugation rates for p-nitrophenol- and 4-methylumbelliferone-glucuronides were at pH 5.1 and for phenolphthalein-glucuronide at pH 6.5. Only conjugation reactions showed latency; the corresponding deconjugation reactions were not latent. UDPglucuronyltransferase is a group of oligomeric isoenzymes with different molecular masses. The molecular masses of the isoenzyme species catalysing the forward and reverse reactions were determined by radiation-inactivation analysis. The molecular masses of the isoenzyme species mediating the catalyses of deconjugation reactions were significantly smaller than those mediating catalyses of conjugation reactions: 66 +/- 4 kDa vs. 109 +/- 7 kDa for p-nitrophenol; 82 +/- 8 kDa vs. 105 +/- 6 kDa for 4-methylumbelliferone; and 74 +/- 8 kDa vs. 159 +/- 14 kDa for phenolphthalein. This suggests that for catalyses of deconjugation reactions only part of a UDPglucuronyltransferase isoenzyme is needed, whereas for forward reactions the complete isoenzymes are required.
材料
货号
品牌
产品描述
Sigma-Aldrich
4-硝基苯基 β-D-葡糖苷酸, ≥98% (TLC)
Sigma-Aldrich
4-硝基苯基 β-D-葡糖苷酸, ≥99.0% (TLC)
Sigma-Aldrich
苯酚 β-D-葡萄糖醛酸, β-glucuronidase substrate
Sigma-Aldrich
苯酚 β-D-葡萄糖醛酸 钠盐, β-glucuronidase substrate