Merck
  • 主页
  • 查找结果
  • Synthesis and characterization of hexa-coordinated Sn(IV) complexes of meso-aryl dipyrrins.
Synthesis and characterization of hexa-coordinated Sn(IV) complexes of meso-aryl dipyrrins.
Dalton transactions (Cambridge, England : 2003) (2013-03-20)
Ritambhara Sharma, Avijit Ghosh, Benedikt Wolfram, Martin Bröring, Mangalampalli Ravikanth
摘要
Our successful synthesis of the first examples of Sn(IV) complexes of meso-aryl dipyrrins is reported. Sn(IV) dipyrrins are synthesized by treating meso-aryl dipyrrins with readily available SnCl2·2H2O in pyridine at reflux temperature and isolated as pure complexes by recrystallization. The X-ray structure revealed that the complex is hexa-coordinated and the Sn(IV) ion is coordinated to two dipyrrin units and two chlorides with overall appearance of a butterfly like structure. The complexes are stable in solution and characterized by spectral and electrochemical techniques.
材料
货号
品牌
产品描述
Sigma-Aldrich
吡啶, anhydrous, 99.8%
Sigma-Aldrich
吡啶, ACS reagent, ≥99.0%
Sigma-Aldrich
吡啶, suitable for HPLC, ≥99.9%
Sigma-Aldrich
吡啶, ReagentPlus®, ≥99%
Sigma-Aldrich
吡啶, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
锡, ≥99%, powder
Sigma-Aldrich
吡啶 盐酸盐, 98%
Sigma-Aldrich
锡, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
吡啶, ≥99%
Sigma-Aldrich
锡, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
锡, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
锡, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Supelco
吡啶, analytical standard
Sigma-Aldrich
锡, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
锡, shot, 99.999% trace metals basis
Sigma-Aldrich
锡, foil, thickness 0.5 mm, 99.998% trace metals basis
Supelco
吡啶 溶液, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
锡, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
吡啶, ACS reagent, ≥99.0%
Sigma-Aldrich
吡啶, biotech. grade, ≥99.9%
Sigma-Aldrich
锡, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
锡, 99.8%, shot, 3 mm
Sigma-Aldrich
吡啶 氢溴酸盐, 98%
Sigma-Aldrich
锡, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
吡啶 盐酸盐, purum, ≥98.0% (AT)
Sigma-Aldrich
吡啶, ReagentPlus®, ≥99%
Supelco
锡(杂质), manufactured by MBH Analytical Ltd (71X SR0)