  • Search Results
  • Characterization of Tyr-Leu-Gly, a novel anxiolytic-like peptide released from bovine αS-casein.
Characterization of Tyr-Leu-Gly, a novel anxiolytic-like peptide released from bovine αS-casein.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2013-04-13)
Takafumi Mizushige, Yurina Sawashi, Ayako Yamada, Ryuhei Kanamoto, Kousaku Ohinata
ABSTRACT
We found previously that dipeptide YL exhibits orally active anxiolytic activity comparable to diazepam. The YL sequence is often observed in the primary structure of natural food proteins. In the present study, we investigated whether YL and YL analogues are released from bovine αS-casein by gastrointestinal proteases. YLG, corresponding to αS1-casein (aa 91-93), was more effectively released from αS-casein than YL by pepsin-pancreatin digestion, mimicking gastrointestinal enzymatic conditions. Using the synthetic model peptide, we determined that trypsin cleaved the N terminus of YLG, and elastase and carboxypeptidase contributed to cleave the C-terminus. YLG exhibited orally active anxiolytic-like activity in the elevated plus maze and open-field tests in mice. The anxiolytic-like activity of YLG was inhibited by WAY100135, SCH23390 or bicuculline, antagonists of serotonin 5-HT1A, dopamine D1, and GABA(A) receptors, respectively; however, YLG had no affinity for these receptors. The pepsin-pancreatin digest of αS-Casein also exhibited anxiolytic-like activity. Meanwhile, anxiolytic-like activity of α-casozepine, an αS1-casein-derived decapeptide with YL sequence in the N terminus, was blocked by WAY100135, SCH23390, or bicuculline, equally to YLG and YL; however, it was not detected in the pepsin-pancreatic digest. Taken together, we found that YLG is released after pepsin-pancreatic digestion of αS-casein and exhibits potent anxiolytic-like activity via activation of serotonin, dopamine, and the GABA receptor system.
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, ≥250 units/mg solid
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
胰脂肪酶 来源于猪胰腺, 8 × USP specifications
Sigma-Aldrich
胰脂肪酶 来源于猪胰腺, 4 × USP specifications
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, ≥400 units/mg protein
Sigma-Aldrich
胰脂肪酶 来源于猪胰腺, powder, suitable for cell culture, 4 × USP specifications
Sigma-Aldrich
胰脂肪酶 来源于猪胰腺, ≥3 × USP specifications
Sigma-Aldrich
(+)-荷包牡丹碱, ≥97.0% (TLC)
胃蛋白酶, powder, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, slightly beige, ≥500 U/mg
Sigma-Aldrich
胃蛋白酶 来源于猪胃粘膜, powder, slightly beige, 1200-2400 U/mg