• 主页
  • 查找结果
  • Elevated levels of DNA-protein crosslinks and micronuclei in peripheral lymphocytes of tannery workers exposed to trivalent chromium.
Elevated levels of DNA-protein crosslinks and micronuclei in peripheral lymphocytes of tannery workers exposed to trivalent chromium.
Mutagenesis (2002-12-11)
M G Medeiros, A S Rodrigues, M C Batoréu, A Laires, J Rueff, A Zhitkovich
摘要
DNA-protein crosslinks (DPC) are a promising biomarker of exposure to hexavalent chromium, a known human carcinogen. Although trivalent chromium is considered to have much lower toxicity, the risk involved in chronic exposure is uncertain. DPC may be a useful tool in clarifying this risk, by signaling an exposure of body tissues to biologically active forms of chromium. DPC quantification was carried out in lymphocytes of a group of tannery workers exposed to trivalent chromium, a small group of manual metal arc stainless steel welders exposed to hexavalent chromium and a control group. This biomarker was compared with the frequency of micronuclei in cytokinesis blocked peripheral lymphocytes as a biomarker of cytogenetic lesions and total plasma and urine chromium levels as an index of exposure. The results indicate a significant increase in the formation of DPC in tannery workers compared with controls (0.88 +/- 0.19 versus 0.57 +/- 0.21%, P < 0.001, Mann-Whitney test) and an even higher level of DPC in welders (2.22 +/- 1.12%, P = 0.03). Tanners showed a significant increase in micronucleated cells compared with controls (6.35 +/- 2.94 versus 3.58 +/- 1.69 per thousand, P < 0.01), whereas in welders this increase was not significant (5.40 +/- 1.67 per thousand ). Urinary chromium was increased in both groups, with a greater increase observed in tanners compared with controls (2.63 +/- 1.62 versus 0.70 +/- 0.38 microg/g creatinine, P < 0.001) than in welders (1.90 +/- 0.37 microg/g creatinine, P < 0.005). Plasma chromium was also increased in both groups (tanners 2.43 +/- 2.11 microg/l, P < 0.001, welders 1.55 +/- 0.67 microg/l, P < 0.005 versus controls 0.41 +/- 0.11 microg/l). In summary, chronic occupational exposure to trivalent chromium can lead to a detectable increase in lymphocyte DNA damage which correlates with a significant exposure of the cells to the metal.
材料
货号
品牌
产品描述
Sigma-Aldrich
铬, chips, 99.995% trace metals basis
Sigma-Aldrich
铬, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
铬, powder, 99.5%, −100 mesh
Sigma-Aldrich
铬, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
铬, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.95% trace metals basis