• 主页
  • 查找结果
  • [The porosity, microhardness, roughness and internal stresses of PMMA base materials in relation to their isolation. 1. Porosity and microhardness].
[The porosity, microhardness, roughness and internal stresses of PMMA base materials in relation to their isolation. 1. Porosity and microhardness].
Zahn-, Mund-, und Kieferheilkunde mit Zentralblatt (1989-01-01)
P Lockowandt, H Loges, I V Wagner
摘要
Selected material properties of hot and cold polymerized denture basic materials (polymethylmethacrylate) were investigated after different methods of isolation: alginate, tin foil and silicone. Both tin foil and silicone isolation the handling of which is much easier result in a decreased portion of porosities and an increased microhardness in the PMMA.
材料
货号
品牌
产品描述
锡, foil, 150x150mm, thickness 0.20mm, as rolled, 98.8%
锡, foil, 0.5m coil, thickness 0.009mm, 97.4%
锡, foil, 100x100mm, thickness 0.125mm, as rolled, 98.8%
锡, foil, 100x100mm, thickness 0.125mm, as rolled, 99.99+%
锡, foil, 100x100mm, thickness 0.15mm, as rolled, 98.8%
锡, foil, 100x100mm, thickness 0.15mm, as rolled, 99.99+%
锡, foil, 100x100mm, thickness 0.1mm, as rolled, 98.8%
锡, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
锡, foil, 100x100mm, thickness 0.20mm, as rolled, 98.8%
锡, foil, 100x100mm, thickness 0.25mm, as rolled, 98.8%
锡, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
锡, foil, 100x100mm, thickness 0.5mm, as rolled, 98.8%
锡, foil, 100x100mm, thickness 0.5mm, as rolled, 99.99+%
锡, foil, 100x100mm, thickness 0.65mm, as rolled, 99.99+%
锡, foil, 100x100mm, thickness 0.75mm, as rolled, 99.99+%
锡, foil, 100x100mm, thickness 1.0mm, as rolled, 98.8%
锡, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99+%
锡, foil, 100x100mm, thickness 1.0mm, as rolled, 99.999%
锡, foil, 100x100mm, thickness 1.5mm, as rolled, 99.99+%
锡, foil, 100x100mm, thickness 2.0mm, as rolled, 98.8%
锡, foil, 100x100mm, thickness 2.0mm, as rolled, 99.99+%
锡, foil, 150x150mm, thickness 0.125mm, as rolled, 98.8%
锡, foil, 150x150mm, thickness 0.125mm, as rolled, 99.99+%
锡, foil, 150x150mm, thickness 0.15mm, as rolled, 98.8%
锡, foil, 150x150mm, thickness 0.1mm, as rolled, 98.8%
锡, foil, 150x150mm, thickness 0.1mm, as rolled, 99.95%
锡, foil, 150x150mm, thickness 0.25mm, as rolled, 98.8%
锡, foil, 150x150mm, thickness 0.25mm, as rolled, 99.99+%
锡, foil, 150x150mm, thickness 0.5mm, as rolled, 98.8%
锡, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99+%
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。